Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rheb
Entrez Gene ID
26954
Gene Name
Ras homolog enriched in brain
Gene Aliases
-
Chromosome Location
Chr.4: 6827429 - 6873384 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013216.1 NP_037348.1 2 - 3 248 111
GenBank mRNA BC088857.1 - 2 - 3 302 111
FQ212847.1 - 2 - 3 270 111
FQ214530.1 - 2 - 3 287 111
FQ218725.1 - 2 - 3 270 111
FQ225701.1 - 2 - 3 252 111
FQ230203.1 - 2 - 3 252 111
U08227.1 - 2 - 3 248 111

View Full Product Details