Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlrp4a
Entrez Gene ID
292545
Gene Name
NLR family, pyrin domain containing 4A
Gene Aliases
RGD1561918
Chromosome Location
Chr.1: 69326303 - 69443748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106220.1 NP_001099690.1 6 - 7 2548 63
XM_008758899.1 XP_008757121.1 7 - 8 2637 63

View Full Product Details