The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106220.1 | NP_001099690.1 | 8 - 9 | 2887 | 134 |
XM_008758899.1 | XP_008757121.1 | 9 - 10 | 2976 | 134 |