The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109321.1 | NP_001102791.1 | 1 - 1 | 3528 | 92 |
XM_006257301.2 | XP_006257363.1 | 5 - 5 | 4190 | 92 | |
XM_006257302.2 | XP_006257364.1 | 6 - 6 | 4140 | 92 | |
XM_006257303.2 | XP_006257365.1 | 5 - 5 | 4470 | 92 | |
XM_006257304.2 | XP_006257366.1 | 4 - 4 | 4413 | 92 | |
XM_006257305.2 | XP_006257367.1 | 5 - 5 | 4086 | 92 | |
GenBank mRNA | BC168654.1 | - | 3 - 3 | 3980 | 92 |