Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slain2
Entrez Gene ID
305310
Gene Name
SLAIN motif family, member 2
Gene Aliases
RGD1311593
Chromosome Location
Chr.14: 37802347 - 37871123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107214.1 NP_001100684.1 1 - 2 629 74
XM_006250898.2 XP_006250960.1 1 - 2 701 74
XM_006250899.2 XP_006250961.1 1 - 2 700 74

View Full Product Details