The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011914.1 | NP_001011914.1 | 2 - 3 | 1004 | 90 |
GenBank mRNA | BC083822.1 | - | 2 - 3 | 1004 | 90 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751317.3 | XP_003751365.1 | - | 1093 | 90 |