Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnfaip3
Entrez Gene ID
683206
Gene Name
tumor necrosis factor, alpha-induced protein 3
Gene Aliases
-
Chromosome Location
Chr.1: 14401103 - 14412822 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748656.3 XP_003748704.2 5 - 6 1106 92

View Full Product Details