Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhlrc2
Entrez Gene ID
307986
Gene Name
NHL repeat containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 277355619 - 277415940 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107444.1 NP_001100914.1 3 - 4 1211 70
XM_006231645.2 XP_006231707.1 3 - 4 1211 70
XM_008760536.1 XP_008758758.1 3 - 4 985 70

View Full Product Details