Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Morn4
Entrez Gene ID
293950
Gene Name
MORN repeat containing 4
Gene Aliases
RGD1307336
Chromosome Location
Chr.1: 261321627 - 261337422 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024975.1 NP_001020146.1 1 - 2 142 69
GenBank mRNA BC091345.1 - 1 - 2 142 69
EF688599.1 - 1 - 2 142 69

View Full Product Details