Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hinfp
Entrez Gene ID
300665
Gene Name
histone H4 transcription factor
Gene Aliases
Mizf, RGD1311492
Chromosome Location
Chr.8: 48626398 - 48638084 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729915.4 XP_002729961.1 6 - 7 1617 68

View Full Product Details