Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Islr
Entrez Gene ID
686539
Gene Name
immunoglobulin superfamily containing leucine-rich repeat
Gene Aliases
-
Chromosome Location
Chr.8: 62946732 - 62949680 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243176.2 XP_006243238.1 1 - 2 240 95

View Full Product Details