Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pih1d2
Entrez Gene ID
315645
Gene Name
PIH1 domain containing 2
Gene Aliases
RGD1310503
Chromosome Location
Chr.8: 55050284 - 55061397 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191793.1 NP_001178722.1 6 - 7 1168 69
XM_006243028.2 XP_006243090.1 5 - 6 934 69
XM_006243029.2 XP_006243091.1 4 - 5 841 69
XM_006243030.2 XP_006243092.1 4 - 5 840 69

View Full Product Details