Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thsd4
Entrez Gene ID
315728
Gene Name
thrombospondin, type I, domain containing 4
Gene Aliases
RGD1566296
Chromosome Location
Chr.8: 64811940 - 65009433 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766376.1 XP_008764598.1 1 - 2 6340 67

View Full Product Details