The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029925.1 | NP_001025096.1 | - | 987 | 71 |
XM_006256577.2 | XP_006256639.1 | 5 - 6 | 1029 | 71 | |
GenBank mRNA | BC097978.1 | - | - | 987 | 71 |