Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Helt
Entrez Gene ID
498633
Gene Name
helt bHLH transcription factor
Gene Aliases
Helt-ps1, Hesl, RGD1564073
Chromosome Location
Chr.16: 49185560 - 49187293 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751600.3 XP_003751648.1 3 - 4 225 75

View Full Product Details