Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galntl6
Entrez Gene ID
361142
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 6
Gene Aliases
Galnt17, RGD1564773
Chromosome Location
Chr.16: 34553966 - 35803016 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135756.1 NP_001129228.1 8 - 9 1303 119
XM_008771227.1 XP_008769449.1 7 - 8 1957 119

View Full Product Details