Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pus7l
Entrez Gene ID
315264
Gene Name
pseudouridylate synthase 7 homolog (S. cerevisiae)-like
Gene Aliases
RGD1304692
Chromosome Location
Chr.7: 135765307 - 135803586 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729847.4 XP_002729893.2 1 - 2 72 61

View Full Product Details