Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jrk
Entrez Gene ID
315073
Gene Name
jerky homolog (mouse)
Gene Aliases
-
Chromosome Location
Chr.7: 115941788 - 115949133 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104612.2 NP_001098082.1 1 - 2 71 96

View Full Product Details