Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc26a10
Entrez Gene ID
366909
Gene Name
solute carrier family 26, member 10
Gene Aliases
RGD1564851
Chromosome Location
Chr.7: 70458528 - 70469099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134595.2 NP_001128067.2 16 - 17 1959 64

View Full Product Details