Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Naga
Entrez Gene ID
315165
Gene Name
N-acetyl galactosaminidase, alpha
Gene Aliases
-
Chromosome Location
Chr.7: 123563047 - 123572104 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012120.1 NP_001012120.1 2 - 3 209 61
XM_006242087.2 XP_006242149.1 3 - 4 423 61
XM_006242088.2 XP_006242150.1 3 - 4 359 61
GenBank mRNA BC082084.1 - 2 - 3 209 61

View Full Product Details