Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parvg
Entrez Gene ID
689069
Gene Name
parvin, gamma
Gene Aliases
-
Chromosome Location
Chr.7: 125288081 - 125307587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130583.1 NP_001124055.1 2 - 3 210 85
XM_006241411.1 XP_006241473.1 2 - 3 167 85
XM_006241412.2 XP_006241474.1 2 - 3 420 85

View Full Product Details