The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244855.1 | NP_001231784.1 | 15 - 16 | 1908 | 57 |
XM_006242100.2 | XP_006242162.1 | 15 - 16 | 3348 | 57 | |
XM_006242101.2 | XP_006242163.1 | 15 - 16 | 2197 | 57 | |
GenBank mRNA | BC082056.1 | - | 15 - 16 | 1816 | 57 |
BP492086.1 | - | 5 - 6 | 459 | 57 | |
FQ217036.1 | - | 15 - 16 | 1892 | 57 |