Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam83f
Entrez Gene ID
315145
Gene Name
family with sequence similarity 83, member F
Gene Aliases
RGD1562089
Chromosome Location
Chr.7: 121889157 - 121918772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130502.1 NP_001123974.1 3 - 4 813 155

View Full Product Details