Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anxa13
Entrez Gene ID
362915
Gene Name
annexin A13
Gene Aliases
-
Chromosome Location
Chr.7: 98217642 - 98270442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134910.1 NP_001128382.1 4 - 5 385 91
XM_008765512.1 XP_008763734.1 4 - 5 717 91

View Full Product Details