Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf19a
Entrez Gene ID
362900
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Gene Aliases
Rnf19
Chromosome Location
Chr.7: 75058758 - 75098331 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130560.1 NP_001124032.1 1 - 2 156 144

View Full Product Details