Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem71
Entrez Gene ID
690657
Gene Name
transmembrane protein 71
Gene Aliases
-
Chromosome Location
Chr.7: 107264235 - 107392808 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271215.1 NP_001258144.1 8 - 9 1055 89

View Full Product Details