Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx31
Entrez Gene ID
366915
Gene Name
sorting nexin 31
Gene Aliases
RGD1309483
Chromosome Location
Chr.7: 75308852 - 75364264 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750338.3 XP_003750386.1 10 - 11 1076 96

View Full Product Details