Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyhr1
Entrez Gene ID
100362155
Gene Name
cysteine/histidine-rich 1
Gene Aliases
-
Chromosome Location
Chr.7: 117706608 - 117719767 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729828.4 XP_002729874.1 - 1016 68

View Full Product Details