Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adsl
Entrez Gene ID
315150
Gene Name
adenylosuccinate lyase
Gene Aliases
null
Chromosome Location
Chr.7: 122157201 - 122192328 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130503.1 NP_001123975.1 1 - 2 191 93

View Full Product Details