Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sdad1
Entrez Gene ID
289504
Gene Name
SDA1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 17247381 - 17274228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006958.1 NP_001006959.1 18 - 19 1649 61
XM_006250722.2 XP_006250784.1 18 - 19 1719 61
GenBank mRNA BC083620.1 - 18 - 19 1649 61

View Full Product Details