Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsph1
Entrez Gene ID
361818
Gene Name
radial spoke head 1 homolog (Chlamydomonas)
Gene Aliases
Tsga2
Chromosome Location
Chr.20: 9998698 - 10020719 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012176.1 NP_001012176.1 - 105 68
GenBank mRNA BC084704.1 - - 105 68

View Full Product Details