Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pnpla1
Entrez Gene ID
361812
Gene Name
patatin-like phospholipase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.20: 6102371 - 6143255 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191841.1 NP_001178770.1 1 - 2 203 68

View Full Product Details