Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spdef
Entrez Gene ID
689210
Gene Name
SAM pointed domain containing ets transcription factor
Gene Aliases
-
Chromosome Location
Chr.20: 7292823 - 7308040 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109530.1 NP_001103000.1 5 - 6 1166 61
XM_006256229.2 XP_006256291.1 5 - 6 1926 61
XM_008772827.1 XP_008771049.1 5 - 6 1036 61
GenBank mRNA BC166744.1 - 5 - 6 1170 61

View Full Product Details