Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ispd
Entrez Gene ID
493574
Gene Name
isoprenoid synthase domain containing
Gene Aliases
Nip
Chromosome Location
Chr.6: 55880133 - 56159466 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008386.1 NP_001008387.1 3 - 4 770 81
XM_008764603.1 XP_008762825.1 3 - 4 2121 81
GenBank mRNA AY769991.1 - 2 - 3 455 81
AY769992.1 - 3 - 4 770 81

View Full Product Details