Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snapc1
Entrez Gene ID
314228
Gene Name
small nuclear RNA activating complex, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.6: 96871675 - 96893422 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108033.1 NP_001101503.1 4 - 5 601 99
GenBank mRNA BC161969.1 - 4 - 5 620 99

View Full Product Details