The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108033.1 | NP_001101503.1 | 9 - 10 | 1140 | 110 |
GenBank mRNA | BC161969.1 | - | 9 - 10 | 1159 | 110 |