Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp2r5e
Entrez Gene ID
299147
Gene Name
protein phosphatase 2, regulatory subunit B', epsilon isoform
Gene Aliases
-
Chromosome Location
Chr.6: 98417974 - 98567017 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106740.1 NP_001100210.1 10 - 11 1626 128
XM_006240222.2 XP_006240284.1 12 - 13 1790 128
XM_006240223.2 XP_006240285.1 12 - 13 1788 128
XM_006240224.2 XP_006240286.1 11 - 12 1528 128
XM_006240225.2 XP_006240287.1 11 - 12 1437 128

View Full Product Details