Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmf1
Entrez Gene ID
681050
Gene Name
polyamine-modulated factor 1
Gene Aliases
-
Chromosome Location
Chr.2: 187752282 - 187771838 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191568.1 NP_001178497.1 3 - 4 360 61

View Full Product Details