Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrnad1
Entrez Gene ID
361976
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
RGD1311265
Chromosome Location
Chr.2: 187294052 - 187302691 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014173.1 NP_001014195.1 4 - 5 863 70
XM_006232686.2 XP_006232748.1 3 - 4 695 70
XM_006232688.2 XP_006232750.1 3 - 4 420 70
XM_006232689.2 XP_006232751.1 3 - 4 695 70
XM_008761179.1 XP_008759401.1 3 - 4 655 70
GenBank mRNA BC079059.1 - 4 - 5 863 70

View Full Product Details