Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrnad1
Entrez Gene ID
361976
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
RGD1311265
Chromosome Location
Chr.2: 187294052 - 187302691 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014173.1 NP_001014195.1 5 - 6 919 67
XM_006232686.2 XP_006232748.1 4 - 5 751 67
XM_006232688.2 XP_006232750.1 4 - 5 476 67
XM_006232689.2 XP_006232751.1 4 - 5 751 67
XM_008761179.1 XP_008759401.1 4 - 5 711 67
GenBank mRNA BC079059.1 - 5 - 6 919 67

View Full Product Details