The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025648.1 | NP_001020819.1 | 1 - 2 | 418 | 71 |
NM_001170576.1 | NP_001164047.1 | 3 - 4 | 335 | 71 | |
GenBank mRNA | BC098864.1 | - | 1 - 2 | 418 | 71 |
DY315040.1 | - | 3 - 4 | 340 | 71 |