Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptx3
Entrez Gene ID
689388
Gene Name
pentraxin 3, long
Gene Aliases
-
Chromosome Location
Chr.2: 158097843 - 158103653 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109536.1 NP_001103006.1 2 - 3 690 78

View Full Product Details