Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc38
Entrez Gene ID
313681
Gene Name
leucine rich repeat containing 38
Gene Aliases
RGD1311416
Chromosome Location
Chr.5: 162031722 - 162053723 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107991.1 NP_001101461.1 1 - 2 1036 94

View Full Product Details