Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wars2
Entrez Gene ID
690654
Gene Name
tryptophanyl tRNA synthetase 2 (mitochondrial)
Gene Aliases
-
Chromosome Location
Chr.2: 201170368 - 201256115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168641.1 NP_001162112.1 2 - 3 357 68

View Full Product Details