Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tlr7
Entrez Gene ID
317468
Gene Name
toll-like receptor 7
Gene Aliases
RGD1563357
Chromosome Location
Chr.X: 28486838 - 28513004 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097582.1 NP_001091051.1 3 - 3 3255 87
XM_006256842.2 XP_006256904.1 2 - 2 3439 87
XM_006256843.2 XP_006256905.1 4 - 4 3530 87
XM_008773170.1 XP_008771392.1 3 - 3 3250 87
GenBank mRNA EF032637.1 - 3 - 3 3255 87

View Full Product Details