The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126092.2 | NP_001119564.1 | 1 - 1 | 218 | 72 |
GenBank mRNA | CB725243.1 | - | 1 - 1 | 218 | 72 |