Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a10
Entrez Gene ID
363229
Gene Name
solute carrier family 39 (zinc transporter), member 10
Gene Aliases
-
Chromosome Location
Chr.9: 59947565 - 60070549 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750698.3 XP_003750746.1 2 - 3 4294 69
XM_006244936.2 XP_006244998.1 2 - 3 1230 69
XM_006244937.2 XP_006244999.1 3 - 4 1218 69
XM_006244938.2 XP_006245000.1 2 - 3 2248 69
XM_008767152.1 XP_008765374.1 2 - 3 1806 69

View Full Product Details