Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap10
Entrez Gene ID
360540
Gene Name
A kinase (PRKA) anchor protein 10
Gene Aliases
D-akap2
Chromosome Location
Chr.10: 48144837 - 48210074 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114606.1 NP_001108078.1 7 - 8 1347 85
XM_006246509.2 XP_006246571.1 7 - 8 2944 85

View Full Product Details