Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a5
Entrez Gene ID
686680
Gene Name
membrane-spanning 4-domains, subfamily A, member 5
Gene Aliases
-
Chromosome Location
Chr.1: 227449923 - 227457762 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749066.3 XP_003749114.1 3 - 4 467 73

View Full Product Details