Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcp10b
Entrez Gene ID
308169
Gene Name
t-complex protein 10b
Gene Aliases
-
Chromosome Location
Chr.1: 53531007 - 53554113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107461.1 NP_001100931.1 5 - 6 850 82

View Full Product Details